ADV412111047 AChemBlock

1,1-Dioxo-tetrahydrothiopyran-4-amine

  • MDL number MFCD09540491
  • Cas Number : 210240-20-3
  • Molecular Weight : 149.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-1-dioxo-tetrahydrothiopyran-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7271
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart