ADV412111050 AChemBlock

(S)-1-Boc-2-(aminomethyl)-4,4-difluoropyrrolidine

  • MDL number MFCD22566249
  • Cas Number : 1363384-67-1
  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-1-boc-2-aminomethyl-4-4-difluoropyrrolidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7274
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart