ADV412111057 AChemBlock

3,5-Dichloropyrazine-2-carbonitrile

  • MDL number MFCD13193460
  • Cas Number : 313339-92-3
  • Molecular Weight : 173.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-5-dichloropyrazine-2-carbonitrile.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7281
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart