ADV412111065 AChemBlock

4-Aminobicyclo[2.2.2]octane-1-carbaxylic acid methyl ester

  • MDL number MFCD20693756
  • Cas Number : 135908-33-7
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-aminobicyclo-2-2-2-octane-1-carbaxylic-acid-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4577
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart