ADV412111067 AChemBlock

1-Boc-3-hydroxypyrrolidine-3-carboxylic acid

  • MDL number MFCD15071383
  • Cas Number : 1067239-08-0
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form orange liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-hydroxypyrrolidine-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7291
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart