ADV412111074 AChemBlock

(2R,4S)-1-Boc-4-methylpyrrolidine-2-carboxylic acid

  • MDL number MFCD23105992
  • Cas Number : 1018818-04-6
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2r-4s-1-boc-4-methylpyrrolidine-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4580
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart