ADV412111079 AChemBlock

6-Chloro-3-oxo-2,3-dihydro-pyridazine-4-carboxylic acid methyl ester

  • MDL number MFCD16250585
  • Cas Number : 89581-64-6
  • Molecular Weight : 188.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-3-oxo-2-3-dihydro-pyridazine-4-carboxylic-acid-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7303
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart