ADV412111084 AChemBlock

Ethyl 5-(methoxymethyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD23105878
  • Cas Number : 1297546-22-5
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-methoxymethyl-1h-pyrazole-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7308
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart