ADV412111085 AChemBlock

1-Boc-6-oxo-1-azaspiro[3.3]heptane

  • MDL number MFCD23105879
  • Cas Number : 1363380-93-1
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-6-oxo-1-azaspiro-3-3-heptane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7309
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart