ADV412111090 AChemBlock

1-Methyl-4-nitro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00464004
  • Cas Number : 4598-86-1
  • Molecular Weight : 171.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-4-nitro-1h-pyrazole-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7314
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart