ADV412111091 AChemBlock

1-Boc-4-iodomethylpiperidine

  • MDL number MFCD08437653
  • Cas Number : 145508-94-7
  • Molecular Weight : 325.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-4-iodomethylpiperidine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7316
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart