ADV412111092 AChemBlock

Ethyl 4-iodopyrazole-3-carboxylate

  • MDL number MFCD23105896
  • Cas Number : 179692-08-1
  • Molecular Weight : 266.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-iodopyrazole-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7317
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart