ADV412111094 AChemBlock

Ethyl trans-1-Boc-4-hydroxypyrrolidine-3-carboxylate

  • MDL number MFCD23105897
  • Cas Number : 849935-83-7
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-trans-1-boc-4-hydroxypyrrolidine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7320
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart