ADV412111096 AChemBlock

(3S,4R)-rel-1-Boc-3-fluoro-4-(hydroxymethyl)piperidine

  • MDL number MFCD24850020
  • Cas Number : 882033-93-4
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3s-4r-rel-1-boc-3-fluoro-4-hydroxymethyl-piperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7322
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart