ADV412111100 AChemBlock

trans-3-Fluorocyclobutanamine hydrochloride

  • MDL number MFCD23106125
  • Cas Number : 1408075-99-9
  • Molecular Weight : 125.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-3-fluorocyclobutanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7326
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart