ADV412111102 AChemBlock

cis-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol

  • MDL number MFCD23105930
  • Cas Number : 1408074-89-4
  • Molecular Weight : 202.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-3-1-1-dimethylethyl-dimethylsilyl-oxy-cyclobutanol.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7328
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart