ADV412111103 AChemBlock

trans-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol

  • MDL number MFCD23105931
  • Cas Number : 1408075-44-4
  • Molecular Weight : 202.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-3-1-1-dimethylethyl-dimethylsilyl-oxy-cyclobutanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7329
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart