ADV412111127 AChemBlock

(3-Fluoroazetidin-3-yl)methanol

  • MDL number MFCD18633069
  • Cas Number : 1268520-93-9
  • Molecular Weight : 105.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White crystal
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-fluoroazetidin-3-yl-methanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7354
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart