ADV412111129 AChemBlock

cis-3-Methoxycyclobutanecarboxylic acid

  • MDL number MFCD22396943
  • Cas Number : 552849-35-1
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-3-methoxycyclobutanecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7356
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart