ADV412111131 AChemBlock

7-Boc-1-oxa-3-oxo-7-azaspiro[3.5]nonane

  • MDL number MFCD22628723
  • Cas Number : 1408076-15-2
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-boc-1-oxa-3-oxo-7-azaspiro-3-5-nonane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7358
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart