ADV412111132 AChemBlock

3-Ethynyloxetan-3-ol

  • MDL number MFCD20444449
  • Cas Number : 1352492-38-6
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-ethynyloxetan-3-ol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7359
pictograms GHS02; GHS07
hstatements H226;H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart