ADV412111137 AChemBlock

tert-Butyl (cis-3-amino-3-methylcyclobutyl)carbamate

  • MDL number MFCD23106027
  • Cas Number : 1408076-24-3
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-cis-3-amino-3-methylcyclobutyl-carbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7364
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart