ADV412111139 AChemBlock

trans-(3-Amino-1-methyl-cyclobutyl)carbamic acid tert-butyl ester

  • MDL number MFCD23106029
  • Cas Number : 1408074-61-2
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-3-amino-1-methyl-cyclobutyl-carbamic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7366
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart