ADV412111140 AChemBlock

(3,3-Difluoro-1-methyl-cyclobutyl)methanol

  • MDL number MFCD23106031
  • Cas Number : 1408076-35-6
  • Molecular Weight : 136.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluoro-1-methyl-cyclobutyl-methanol.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7367
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart