ADV412111144 AChemBlock

(R)-1,4-Diazabicyclo[3.2.1]octane dihydrochloride

  • MDL number MFCD23704632
  • Cas Number : 857334-81-7
  • Molecular Weight : 185.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-1-4-diazabicyclo-3-2-1-octane-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7373
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart