ADV412111155 AChemBlock

2-(Methylthio)-4-methyl-8H-pyrido[2,3-d]pyrimidin-7-one

  • MDL number MFCD18632640
  • Cas Number : 846036-77-9
  • Molecular Weight : 207.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-methylthio-4-methyl-8h-pyrido-2-3-d-pyrimidin-7-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7386
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart