ADV412111160 AChemBlock

1-Boc-3-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-yl]azetidine

  • MDL number MFCD18383285
  • Cas Number : 877399-35-4
  • Molecular Weight : 349.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-pyrazol-1-yl-azetidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7391
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart