ADV412111161 AChemBlock

2-methyl-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]propan-1-ol

  • MDL number MFCD22571853
  • Cas Number : 1040377-18-1
  • Molecular Weight : 266.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazole-1-ethanol-beta-beta-dimethyl-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7392
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart