ADV412111202 AChemBlock

(2R,5S)-1,2,5-Trimethylpiperazine oxalate

  • MDL number MFCD26959165
  • Cas Number : 1523541-95-8
  • Molecular Weight : 218.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2r-5s-1-2-5-trimethylpiperazine-oxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7465
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart