ADV412111207 AChemBlock

3-Benzyloxy-6-(tert-butyl-dimethylsilanyloxymethyl)-4-oxo-4H-pyran-2-carboxylic acid

  • MDL number MFCD26959170
  • Cas Number : 895134-29-9
  • Molecular Weight : 390.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-benzyloxy-6-tert-butyl-dimethylsilanyloxymethyl-4-oxo-4h-pyran-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7479
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart