ADV412111210 AChemBlock

8-Oxa-bicyclo[3.2.1]octane-3-carbonitrile

  • MDL number MFCD26959172
  • Cas Number : 855386-64-0
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-oxa-bicyclo-3-2-1-octane-3-carbonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7512
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart