ADV412111219 AChemBlock

(S)-2-N-Boc-aminomethylmorpholine oxalate

  • MDL number MFCD26959181
  • Cas Number : 1820575-28-7
  • Molecular Weight : 306.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-2-n-boc-aminomethylmorpholine-oxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1124
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart