ADV412111229 AChemBlock

(3-tert-Butoxycarbonylamino-oxetan-3-yl)acetic acid

  • MDL number MFCD16140246
  • Cas Number : 1221715-78-1
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-tert-butoxycarbonylamino-oxetan-3-yl-acetic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7429
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart