ADV412111236 AChemBlock

3-(4-Fluorophenyl)oxetane-3-carboxylic acid

  • MDL number MFCD22544051
  • Cas Number : 1393572-06-9
  • Molecular Weight : 196.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-4-fluorophenyl-oxetane-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7436
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart