ADV412111242 AChemBlock

3-(4-Bromophenyl)-3-methyl-oxetane

  • MDL number MFCD26393310
  • Cas Number : 872882-97-8
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-4-bromophenyl-3-methyl-oxetane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7443
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart