ADV412111265 AChemBlock

Octahydropyrazino[2,1-c][1,4]oxazine dihydrochloride

  • MDL number MFCD19442255
  • Cas Number : 1257998-65-4
  • Molecular Weight : 215.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/octahydropyrazino-2-1-c-1-4-oxazine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7469
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart