ADV412111272 AChemBlock

N,N-Dimethyl-2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]ethanamine

  • MDL number MFCD16660235
  • Cas Number : 877149-80-9
  • Molecular Weight : 265.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-n-dimethyl-2-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-ethanamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7476
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart