ADV412111276 AChemBlock

2-Amino-4-chloro-5-methylpyrimidine

  • MDL number MFCD00725898
  • Cas Number : 20090-58-8
  • Molecular Weight : 143.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-4-chloro-5-methylpyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7406
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart