ADV412111300 AChemBlock

9-Boc-3-oxo-2,9-diaza-spiro[5.5]undecane

  • MDL number MFCD17017451
  • Cas Number : 1251021-18-7
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/9-boc-3-oxo-2-9-diaza-spiro-5-5-undecane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7407
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart