ADV412111304 AChemBlock

2-Bromo-5,8-dioxaspiro[3.4]octane

  • MDL number MFCD20637976
  • Cas Number : 1257996-82-9
  • Molecular Weight : 193.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-5-8-dioxaspiro-3-4-octane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7409
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart