ADV412111310 AChemBlock

Methyl trans-4-(Boc-amino)cyclohexanecarboxylate

  • MDL number MFCD17015041
  • Cas Number : 146307-51-9
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-trans-4-boc-amino-cyclohexanecarboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7520
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart