ADV412111319 AChemBlock

1,3-Dimethyl-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD07625944
  • Cas Number : 197367-87-6
  • Molecular Weight : 184.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-3-dimethyl-1h-pyrazol-4-amine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-858-1
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart