ADV412111326 AChemBlock

trans-3-Methoxycyclobutanamine hydrochloride

  • MDL number MFCD22415180
  • Cas Number : 1408074-49-6
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-3-methoxycyclobutanamine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3622-1
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart