ADV412111330 AChemBlock

trans-3-(Aminomethyl)cyclobutanol hydrochloride

  • MDL number MFCD22422286
  • Cas Number : 1404365-04-3
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-3-aminomethyl-cyclobutanol-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2432-01
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart