ADV412111334 AChemBlock

(2R,4R)-Benzyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD18837818
  • Cas Number : 1009335-39-0
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2r-4r-benzyl-4-hydroxy-2-hydroxymethyl-pyrrolidine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-536
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart