ADV412111342 AChemBlock

(S)-1-Benzyl-2-methylpiperazine

  • MDL number MFCD08059564
  • Cas Number : 511254-92-5
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-1-benzyl-2-methylpiperazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-547
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart