ADV412111345 AChemBlock

(S)-Benzyl 2-methylpiperazine-1-carboxylate

  • MDL number MFCD03840851
  • Cas Number : 923565-98-4
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-benzyl-2-methylpiperazine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-550
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart