ADV412111357 AChemBlock

tert-butyl 7-benzyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate

  • MDL number MFCD19443918
  • Cas Number : 227940-71-8
  • Molecular Weight : 316.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-7-diazabicyclo-3-3-1-nonane-3-carboxylic-acid-7-phenylmethyl-1-1-dimethylethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-564
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart