ADV412111380 AChemBlock

tert-Butyl 3,7-diazabicyclo[3.3.1]nonane-3-carboxylate

  • MDL number MFCD17016673
  • Cas Number : 227940-72-9
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-7-diazabicyclo-3-3-1-nonane-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P264;P280;P301+312;P302+P352;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-589
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart