ADV412111385 AChemBlock

Thiomorpholine-3-carboxylic acid hydrochloride

  • MDL number MFCD08436167
  • Cas Number : 96612-95-2
  • Molecular Weight : 183.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/thiomorpholine-3-carboxylic-acid-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P264;P280;P301+312;P302+P352;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-595
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart